Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Polypropylene Laboratory Tray
SDPSCI_ED

Trays that can be used for multiple different applications.

$151.00 - $235.00

Specifications

Color White
Height (Metric) 76.2 mm
Material Polypropylene
Type Laboratory Tray
Height (English) 3 in.
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W x H) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W x H) Price Quantity & Availability  
S01237
SDPSCI_ED
View Documents
United Scientific™
81701
18 x 14 x 3 in.
Pack of 10 for $235.00
Add to Cart
 
S01238
SDPSCI_ED
View Documents
United Scientific™
81702
15 x 12 x 3 in.
Pack of 10 for $151.00
Add to Cart
 
Description

Description

  • Multi-purpose polypropylene molded trays can be used for sterilizing, drying glassware, porcelain ware, and other laboratory supplies
  • These steam autoclavable trays have tapered walls designed for nesting
Specifications

Specifications

White
Polypropylene
3 in.
76.2 mm
Laboratory Tray
Autoclavable
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit