Learn More
Ops Diagnostics Polypropylene 2 mL Disruption Unskirted Tubes & Caps, 500/cs
Supplier: Ops Diagnostics PPDT 0250014
Polypropylene 2 mL disruption tubes with o-ring cap, clear. Tubes are autoclavable up to 121°C. Case of 500.