Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform™ Polyethylene Safety Sign, DANGER HIGH VOLTAGE
Signs identify and indicate the potential, type, and degree of hazards that may lead to accidental injury or property damage. They comply in accordance with OSHA 29 CFR 1910.145 and ANSI Z535 requirements.
$11.47 - $16.57
Specifications
Material | Polyethylene |
---|---|
Mounting Type | Corner Holes |
Shape | Rectangular |
Languages | English |
Displayed Text | DANGER HIGH VOLTAGE |
Catalog Number | Mfr. No. | Material | Mounting Type | Size (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Mounting Type | Size (English) | Price | Quantity & Availability | |||
19-111-139
|
Accuform™
MELC113VP |
Polyethylene | Corner Holes | 7 x 10 in. |
Each for $11.47
|
|
|||
19-111-140
|
Accuform™
MELC114VP |
Polyethylene | Corner Holes | 10 x 14 in. |
Each for $16.57
|
|
|||
Description
- Durable plastic has slight flex that ideal for indoor and sheltered outdoor environments
- Printed surface has clear UV protective overcoat
- 50% recycled content with UV inhibitors and good impact resistance
- Rounded corners with 3/16 in. mounting holes for installation
Specifications
Polyethylene | |
Rectangular | |
DANGER HIGH VOLTAGE |
Corner Holes | |
English | |
Red on White |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title