Learn More
Polysciences, Inc. Poly/Bed 812 (Luft formulations) Embedding Kit/DMP-30
Supplier: Polysciences, Inc. 087921
Use Poly/Bed for your scientific studies requiring beam stability, low shrinkage, and good contrast. Poly/Bed Epoxy formulations are of lower viscosity than Araldite formulations, and hence provide better penetration into tissue samples.