Learn More
Tocris Bioscience™ Polyamine Supplement x1000 (lyophilized)
Media supplement to boost cell growth; used as component of CEPT cocktail to enhance stem cell viability
Supplier:  Tocris Bioscience™ 7739/1 PACK
Catalog No. 77391PACK
Add to Cart
This item is not returnable. View return policy
Description
Description
Polyamine Supplement x1000 (lyophilized) is a media supplement comprising a lyophilized mixture of Putrescine, Spermine (Cat. No. 0958) and Spermidine. The product is used as a component of the CEPT cocktail, in combination with Emricasan (Cat. No. 7310), Chroman 1 (Cat. No. 7163) and trans-ISRIB (Cat. No. 5284) to enhance viability of human pluripotent stem cells (hPSCs) and their differentiated progeny. Polyamine Supplement x1000 (lyophilized) also improves survival of pluripotent and differentiated cells during cryopreservation, organoid formation, single-cell cloning, and genome editing.
NOTE: This reagent is provided lyophilized. To use prepare a 1000x stock solution by reconstituting vial contents in 5 mL of ddH2O and filter sterilize before use. Add 1 mL of sterile resuspended polyamines supplement (1000x stock) per 1 L of media.
Specifications
Specifications
1 Pk. (1 x Vial)
Compounds for Induced Pluripotent Stem Cells
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
Recommended Storage : Store at -20°C