Learn More
Azzota Corp BLUE POLYPROPYLENE SCREW THREAD CLOSURES CAPS, PTFE/SILICONE SEPTUM, Red PTFE, White Silicone Septum, 1000/pk
Supplier: Azzota Corp C104890051000RED
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
BLUE POLYPROPYLENE SCREW THREAD CLOSURES CAPS, PTFE/SILICONE SEPTUM, Red PTFE, White Silicone Septum, 1000/pk.