Learn More
Invivogen POLY(I:C) (HMW) VACCIGRADE
Supplier: Invivogen VACPIC
Poly(I:C) is a synthetic double-stranded RNA, that can activate the immune response through two distinct PRRs. Endosomal poly(I:C) activates TLR3 while cytosolic poly(I:C) activates RIG-I/MDA-5. Triggering the TLR3 pathway induces IL-12 and type I IFNs production, while stimulation of MDA-5 enhances the production of type I IFNs. Poly(I:C) promotes Th1 biased immunity through its induction of IL-12 and type I IFNs.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.