Learn More
Azzota Corp Polarimeter Tube, Cap Model, High Clear Borosilicate Glass, Diameter: 14mm, Pathlength: 50mm Or 100mm Or 200mm
Supplier:  Azzota Corp APTC200
Encompass_Preferred
Polarimeter tube, cap model, High Clear Borosilicate Glass, Diameter: 14mm, Pathlength: 50mm or 100mm or 200mm
Catalog No. 50-753-2956