Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Lactate Liquid Reagents
Quantify lactate levels in plasma
Supplier: Pointe Scientific L759650
Includes:
- Reagent 1: TRIS Buffer 100mM; 4-aminoantipyrene 1.7mM; Peroxidase (Horseradish) >10,000 U/L; Surfactant; Stabilizer; Sodium azide 0.09%
- Reagent 2: TRIS Buffer 100mM; Lactate Oxidase (Microbial) >1000 U/L; TOOS 1.5mM; Surfactant; Stabilizer; Sodium azide 0.09%
Requires : Lactate standard or suitable serum-based calibrator
This item is not returnable. View return policy
Description
- Method: Lactate Oxidase, Endpoint
- Format: Liquid (ready to use)
- Wavelength: 546/700nm
- Linearity: 0 to 15mM
- Expected Values: 0.5 to 2.2mM
Applications:
For the quantitative determination of lactate in human plasma
Specifications
CE Marked | |
Tris Buffer 100mM, 4-aminoantipyrene 1.7mM, Peroxidase (Horseradish) >10000U/L, Surfactant, Stabilizer, Sodium Azide 0.09%, Lactate Reagent (R2): Tris Buffer 100mM, Lactate Oxidase (Microbial) >1000U/L, TOOS 1.5mM, Surfactant, Stabilizer Sodium Azide 0.09% as preservative | |
Lactate R1, Lactate R2 | |
546/700 nm |
Lactate Set | |
Quantitative determination of Lactate in Human Plasma | |
1 x 30 mL, 1 x 20 mL | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For in vitro diagnostic use only
Spot an opportunity for improvement?
Provide Content Correction