Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific CK (Liquid) Reagents
$378.00 - $1205.00
Specifications
Type | Creatine Kinase Reagent Kit |
---|---|
Formulation | CK R1 (Buffer) contains: Imidazole Buffer 100mM/L (pH 6.7), NADP 2mM/L, HK (Baker's Yeast) 2.5KU/L, Glucose 20mM/L, Magnesium Acetate 10mM/L, EDTA 2mM/L, N-acetylcysteine (NAC) 20mM/L, CK R2 (enzyme reagent) contains: Imidazole Buffer 100mM/L (pH 6.7), A |
For Use With (Application) | Quantitative determination of Creatine kinase activity in Serum |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-666-208
|
Pointe Scientific
C7522150 |
150 mL |
Each for $378.00
|
|
|||||
23-666-209
|
Pointe Scientific
C7522450 |
450 mL |
Each for $995.00
|
|
|||||
23-666-210
|
Pointe Scientific
C7522625 |
625 mL |
Each for $1,205.00
|
|
|||||
Specifications
Creatine Kinase Reagent Kit | |
CK R1 (Buffer) contains: Imidazole Buffer 100mM/L (pH 6.7), NADP 2mM/L, HK (Baker's Yeast) 2.5KU/L, Glucose 20mM/L, Magnesium Acetate 10mM/L, EDTA 2mM/L, N-acetylcysteine (NAC) 20mM/L, CK R2 (enzyme reagent) contains: Imidazole Buffer 100mM/L (pH 6.7), A | |
Quantitative determination of Creatine kinase activity in Serum | |
Liquid |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title