Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Cholesterol Liquid Reagents
$133.00 - $757.00
Specifications
Certifications/Compliance | CE Marked |
---|---|
Type | Cholesterol (Liquid) Reagent Kit |
Formulation | 4-Aminoantipyrine 0.25mM, Cholesterol Esterase >150U/L, Cholesterol Oxidase >150U/L, Peroxidase >1500U/L, Phenol >15mM, pH 6.8 Phosphate Buffer, non-reactive Stabilizers and preservatives |
For Use With (Application) | Quantitative determination of total Cholesterol in Serum |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-666-200
|
Pointe Scientific
C7510120 |
1 x 120 mL |
Each for $133.00
|
|
|||||
23-666-202
|
Pointe Scientific
C7510500 |
1 x 500 mL |
Each for $462.00
|
|
|||||
23-666-201
|
Pointe Scientific
C75101L |
1 x 1 L |
Each for $757.00
|
|
|||||
Specifications
CE Marked | |
4-Aminoantipyrine 0.25mM, Cholesterol Esterase >150U/L, Cholesterol Oxidase >150U/L, Peroxidase >1500U/L, Phenol >15mM, pH 6.8 Phosphate Buffer, non-reactive Stabilizers and preservatives | |
Liquid |
Cholesterol (Liquid) Reagent Kit | |
Quantitative determination of total Cholesterol in Serum |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title