Learn More
Pointe Scientific Bilirubin Total Reagents for Hitachi Analyzers
Supplier: Pointe Scientific HB979693
This item is not returnable.
View return policy
Specifications
CE Marked | |
Total Bilirubin R1 Acid Buffer 50mM/L, Surfactants, Total Bilirubin R2 Acid Buffer >30mM/L, DPD >2mM/L and Stabilizers | |
693 mL |
Total Bilirubin Reagent Kit | |
Quantitative determination of total Bilirubin in Serum, in-vitro diagnostic use only | |
Liquid |