Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Auto LDL Cholesterol Liquid Reagents
For direct quantitative determination of low density lipoprotein cholesterol (LDL-C) in human serum or plasma
Supplier: Pointe Scientific L757440
This item is not returnable.
View return policy
Description
Requires autoHDL/LDL Calibrator (23-666-140).
- Method: Direct/Automated
- Format: Liquid
- Wavelength: 546/660nm
- Linearity: 0-700mg/dL
- Expected Values: >130mg/dL
- Reconstituted Stability: Ready to use liquid
Specifications
CE Marked | |
R1 contains: MES Buffer pH 6.3, Detergent 1, Cholesterol Esterase, Cholesterol Oxidase, Peroxidase, 4-aminoantipyrine, Ascorbic Acid Oxidase, Preservative, R2 contains: MES Buffer pH 6.3, Detergent 2, N,N-bis (4-sulfhobutyl)-m Toluidine-disodium (DSBmT), | |
1 x 30mL R1, 1 x 10mL R2 |
AutoLDL™ Cholesterol Reagent Kit | |
Quantitative determination of Low Density Lipoprotein Cholesterol in Human Serum or Plasma | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction