Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Amylase Reagents
Test α-amylase activity for diagnoses
$286.00 - $2740.00
Specifications
Certifications/Compliance | CE Marked |
---|---|
Type | Amylase Reagent |
Formulation | MES Buffer pH 6, 2-Chloro-p-Nitrophenyl-alpha-D-Maltotrioside 1.8mM, Sodium Chloride 350mM, Calcium Acetate 6mM, Potassium Thiocyanate 900mM, Sodium Azide 0.1% |
For Use With (Application) | Quantitative kinetic determination of a-amylase activity in Human Serum |
Wavelength | 405 nm |
Includes: MES Buffer pH 6.0±0.1; 2-Chloro-p-Nitrophenyl-α-D-Maltotrioside1.8mM; Sodium Chloride 350mM; Calcium Acetate 6mM; Potassium Thiocyanate 900mM; Sodium Azide 0.1%
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-666-111
|
Pointe Scientific
A756460 |
2 x 30 mL |
Pack of 2 for $286.00
|
|
|||||
23-666-113
|
Pointe Scientific
A7564120 |
4 x 30 mL |
Pack of 4 for $522.00
|
|
|||||
23-666-112
|
Pointe Scientific
A7564480 |
4 x 120 mL |
Pack of 4 for $1,566.00
|
|
|||||
23-666-110
|
Pointe Scientific
A75641L |
1 x 1 L |
Each for $2,740.00
|
|
|||||
Description
- Method: Kinetic, CNP3G
- Format: Liquid (ready to use)
- Wavelength: 405nm
- Linearity: 0 to 2000U/L
- Expected Values: 25 to 125U/L
For quantitative kinetic determination of α-amylase activity in human serum. Most commonly used for diagnosis and treatment of pancreatic diseases.
Specifications
CE Marked | |
MES Buffer pH 6, 2-Chloro-p-Nitrophenyl-alpha-D-Maltotrioside 1.8mM, Sodium Chloride 350mM, Calcium Acetate 6mM, Potassium Thiocyanate 900mM, Sodium Azide 0.1% | |
405 nm |
Amylase Reagent | |
Quantitative kinetic determination of a-amylase activity in Human Serum | |
Liquid |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title