Learn More
Pluriselect pluriStrainer Mini (Cell Strainer) 20 um mesh-size, 100 pcs. (4x25), sterile
Supplier: Pluriselect 431002060
The pluriStrainer Mini is a sieving device (or cell strainer) for a wide range of laboratory applications where filtration and purification of liquids is required. It has been optimized for small sample volumes upto 700 ul. Its unique design allows fitting on a wide variation of tubes, e.g. 1.5 ml /2.0 ml reaction tubes, 15 ml conical centrifuge tubes, FACS-tubes, Cryo vials, 24 well plate and 48 well plate.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.