Learn More
Chemglass Life Sciences Plug, Drosophila, High Density Foam, for Narrow Vials
SDP
Supplier:  Chemglass Life Sciences CGE4790001
Encompass_Preferred
• Fly plugs are made of high-density foam• Causes minimal shedding and reduce evaporation• Maintains tight fit after repeated use and removal• Plugs are compatible with CGE-4789 Drosophila vials and bottles (Sold Separately)
Catalog No. 50-189-7636
Explore available promotions