Learn More
Cell Biolabs Inc pLenti-GFP Lentiviral Control Vector
Supplier: Cell Biolabs Inc LTV400
The pLenti-GFP Lentiviral Control Vector may be co-transfected with third-generation lentiviral packaging constructs to make a lentiviral particle containing GFP. The resulting lentivirus is useful as a transduction control.