Learn More
Perkin Elmer US LLC High Temperature Platinum Sample Pan Kit
Supplier: Perkin Elmer US LLC N5190280
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The High Temperature Platinum Sample Pan Kit includes four platinum sample pans for high temperature analysis on the TGA 7 and PYTIS 1 TGA analyzers.