Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Platinum Cobalt Color Standard APHA 500), Standard, Macron Fine Chemicals™
macron
$5845.00 - $7747.00
Specifications
Chemical Name or Material | Platinum Cobalt Color Standard (APHA 500) |
---|---|
Physical Form | Solution |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MK-H296-100
|
Avantor
H29601 |
100 mL |
Case of 12 for $5,845.00
|
|
|||||
MK-H296-500
|
Avantor
H29603 |
500 mL |
Case of 12 for $7,747.00
|
|
|||||
Specifications
Platinum Cobalt Color Standard (APHA 500) | |
Solution |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title