Learn More
Medicus Health Wire Media Plate Holder For 48 Petri Dishes
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Medicus Health 5310M1
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
∙ Lightweight and rust free Stainless Steel Media Holders have a capacity of 48 Petri Dishes ∙These higher capacity racks hold up to 48 culture plates and will improve workflow and organization ∙Wire racks can be safely stacked with nesting tabs to maximize incubator space ∙Racks are suitable for storing, incubating, sorting and finally autoclaving media plates ∙Wire design lets you clearly visualize culture dishes ∙Caddy & Wire Racks are fully autoclavable∙Optional tote holds two Wire Media Racks and is available as an accessory or can be ordered in a combination package ∙Overall Size: 12-1/2” L x 4-1/2” W x 9” H
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.