Learn More
Fine Science Tools Instrument Cases, Yellow, 17 x 8 x 2 cm
SDP
Supplier:  Fine Science Tools 2083003
Encompass
Product: Instrument Cases - 17 x 8 x 2cm/ Dimensions: 17 x 8 x 2cm/ Alloy / Material: Plastic/ Color: Yellow/ Autoclave Safe: Yes/ Unit: Each
Catalog No. NC1137204