Learn More
United Scientific Supplies Plastic Clamps for Jointed Glassware
For use with 24/40 joint size glassware
Supplier: United Scientific Supplies CLJP24
This item is not returnable.
View return policy
Description
- Plastic conical joint clips are size 24
- Designed to securely hold 24/40 joint size glassware together
Specifications
Glassware Clip | |
Plastic | |
1 Ea. |
24/40 | |
24/40 Jointed Glassware |