Learn More
Uline Plastic Drum - 15 Gallon, Closed Top, Natural
Supplier: Uline S19418
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Excellent for indoor or outdoor use. Corrosion free, dent-resistant polyethylene. 15 gallon stores up to 230 lbs. of liquids and solids. Two openings to pour and vent: 2" and 3/4" diameter. FDA compliant resins.