Learn More
Invivogen Plasmocure™ Antimycoplasma agent, 100 mg (1 ml)
SDP
Supplier:  Invivogen ANTPC
Encompass
Plasmocure: Mycoplasma Elimination Reagent. Product concentration: 100 mg/ml, Sterile ready-to-use solution, Elimination of mycoplasmas in two weeks (recommended concentration of 50 i1/4g/ml), Shipped at room temperature. Unit size 100 mg (1 ml)
Catalog No. NC9992369