Learn More
Grainger BEING SCIENTIFIC, Bath, 5 L Capacity
Supplier:  Grainger 60HU96
Encompass_Preferred
The Being Scientific Bath 5L Capacity is a 5 liter plastic beaker made from polypropylene with a pour spout and handle for easy pouring. It has volume markings up to 5 liters and is autoclavable up to 121°C. The beaker has a wide mouth opening and can be used for mixing and storage. It is lightweight, durable, and chemical resistant. The dimensions are 18cm diameter x 24cm height and it has a capacity of 5000ml.
Catalog No. NC2597738