Learn More
Eppendorf™ Pipette Manager PROMO
Pipette manager allows you to easily transfer volumes to connected electronic pipettes from Eppendorf™ and more.
Supplier:  Eppendorf™ 2231000956
Catalog No. 02-911-585PMO
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
  • An external touch server enabling the communication with connected Eppendorf™ Xplorer electronic pipettes
  • Easily enter pipetting volumes and other parameters using the touch panel
  • Settings are immediately transferred to connected electronic pipettes, allowing you to start working right away
  • Get guidance from the software-embedded Liquid Manager when working with liquids of different viscosity, volatility or foam-building liquids
  • Choose from predefined Eppendorf™ liquid types (e.g. acetone) or create your own
  • Automatically document pipetting activities via Pipetting Records and easily export your recordings as PDF file via USB
  • Manage your pipette fleet more effectively within your working group and enhance collaboration (e.g. through sharing created liquid types)
  • All Eppendorf™ Xplorer and Xplorer plus 4, 6, 8 and 12-channel electronic pipettes are compatible
    • Move It™ pipettes are currently not supported
  • Easily upgrade Eppendorf™ Xplorer and Xplorer plus electronic pipettes with the Eppendorf™ Xplorer connect Wi-Fi module
    • Wi-Fi modules comes with shorter battery for an optimal fit
TRUSTED_SUSTAINABILITY
Specifications
Specifications
External Touch Server
Product Suggestions
Product Suggestions