Learn More
Hach Company Pipet, Volumetric, Class A, 50 mL
Supplier: Hach Company 1451541
Glass Class A pipets unserialized and calibrated “To Deliver”. Designed to ASTM E969 specification for accuracy and ASTM E1273 for color coding.