Learn More
Wilmad Labglass PIPET VOL CLASS B 50ML
Supplier: Wilmad Labglass LG9355132
Volumetric Pipet, Class 'B', To Deliver, Color-Coded. Capacity: 50mL. Capacity Tolerance: 0.10mL. Color Band: Red. Manufactured to meet the requirements for accuracy, appearance, glass quality and calibration line accuracy of ASTM E969.