Learn More
Wilmad Labglass PIPET VOL CLASS B 20ML
Supplier: Wilmad Labglass LG9355124
Volumetric Pipet, Class 'B', To Deliver, Color-Coded. Capacity: 20mL. Capacity Tolerance: 0.06mL. Color Band: Yellow. Manufactured to meet the requirements for accuracy, appearance, glass quality and calibration line accuracy of ASTM E969.