Learn More
Wilmad Labglass PIPET VOL CLASS A 2ML
Supplier: Wilmad Labglass LG9350102
Volumetric Pipet, Class 'A', To Deliver, Color-Coded. Capacity: 2mL. Capacity Tolerance: 0.006mL. Color Band: Orange. Manufactured to meet the requirements for accuracy, appearance, glass quality and calibration line accuracy of ASTM E969.