Learn More
Wilmad Labglass PIPET VOL CLASS A 200ML
Supplier: Wilmad Labglass LG9350136
Volumetric Pipet, Class 'A', To Deliver, Color-Coded. Capacity: 200mL. Capacity Tolerance: 0.10mL. Color Band: Blue. Manufactured to meet the requirements for accuracy, appearance, glass quality and calibration line accuracy of ASTM E969.