Learn More
Wilmad Labglass PIPET VOL CLASS A 0.5ML
Supplier: Wilmad Labglass LG9350100
Volumetric Pipet, Class 'A', To Deliver, Color-Coded. Capacity: 0.5mL. Capacity Tolerance: 0.006mL. Color Band: 2 Black. Manufactured to meet the requirements for accuracy,appearance,glass quality and calibration line accuracy of ASTM E969.