Learn More
Wilmad Labglass PIPET SERO CL B 5ML BLUE BD
Supplier: Wilmad Labglass LG9335116
Serological Pipet, Class 'B', To Deliver, Color-Coded. Capacity: 5mL. Subdivision: 0.1mL. Capacity Tolerance: 0.04mL. Color Band: Blue. Calibrated to deliver at 20degreeC. Pipet has permanent amber scale.