Learn More
Wilmad Labglass PIPET SERO CL B 2ML GREEN BD
Supplier: Wilmad Labglass LG9335112
Serological Pipet, Class 'B', To Deliver, Color-Coded. Capacity: 2mL. Subdivision: 0.1mL. Capacity Tolerance: 0.02mL. Color Band: Green. Calibrated to deliver at 20degreeC. Pipet has permanent amber scale.