Learn More
Wilmad Labglass PIPET SERO CL B 25ML WHT BD
Supplier: Wilmad Labglass LG9335120
Serological Pipet, Class 'B', To Deliver, Color-Coded. Capacity: 25mL. Subdivision: 0.1mL. Capacity Tolerance: 0.10mL. Color Band: White. Calibrated to deliver at 20degreeC. Pipet has permanent amber scale.