Learn More
Wilmad Labglass PIPET SERO CL B 1ML YELLOW BD
Supplier: Wilmad Labglass LG9335110
Serological Pipet, Class 'B', To Deliver, Color-Coded. Capacity: 1mL. Subdivision: 0.01mL. Capacity Tolerance: 0.02mL. Color Band: Yellow. Calibrated to deliver at 20degreeC. Pipet has permanent amber scale.