Learn More
PIP™ The Scout™ Rimless Safety Glasses with Clear Temple and Clear Lens
Click to view available options
No. per Pack:
1 Ea.
12/Pk.
Description
- Polycarbonate unilens
- Over-the-glass design fits over all prescription eyewear
- Can also be worn by itself as a regular safety glass
- Molded nose bridge design to fit the vast majority of users
- Indirect vent temples to reduce fogging
- Clear Lens - clear, crisp and distortion free visibility while meeting the requirement standards that help provide protection from projectiles and impact in general use applications
- Uncoated
- Economical style intended to be used by visitors to a facility, or other short-term applications
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
Specifications
Lens Tint | Clear |
No. per Pack | 1 Ea. |
Temple Style | Vented |
Lens Material | Polycarbonate |