Learn More
Electron Microscopy Sciences Cambridge SEM Pin Stub Adapter 500 ML
Supplier: Electron Microscopy Sciences EMS002
This adapter will convert 1/8" (3.1 mm) pin stub holders to the re-entrant type 1 1/4" (32 mm).