Learn More
Fine Science Tools PINS ELEFANT INSECT
Supplier: Fine Science Tools 2600065
Insect Pins - Black Anodized/Size 6/ Size: 6/ Tip Diameter: 0.03mm/ Rod Diameter: 0.65mm/ Alloy: Black Anodized Steel/ Length: 4cm/ Unit: 100