Learn More
Epredia™ Pinch-Opening White Biopsy Bags
Feature an easy to pinch opening that allows for quick specimen insertion while still maintaining sample security. Biopsy Bags are made from solvent-resistant nylon and conveniently packaged, designed to grab and go.
Catalog Number | Mfr. No. | Size | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Quantity | Price | Quantity & Availability | ||||
67-740-14W
|
Epredia™
6774014W |
Small | 200/Cs. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
67-740-10W
|
Epredia™
6774010W |
Small | 1000/Cs. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Minimize the risk of small specimen loss
- Reduce reagent carryover and pressure artifact often found with sponges
- Rapidly remove specimen with convenient peel-open bag
Specifications
Biopsy Bag | |
Nylon | |
White |