Learn More
TARGETMOL CHEMICALS INC PIN1 INHIBITOR API-1 5MG
Supplier: TARGETMOL CHEMICALS INC T16538
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PIN1 inhibitor API-1 is a specific Pin1 inhibitor (IC50 72.3 nM). PIN1 inhibitor API-1 retains the active conformation of pXPO5 and restores the ability of pXPO5 to transport pre-miRNAs from the nucleus to cytoplasm thus up-regulating the anticancer miRNA biogenesis to suppress both in vitro and in vivo hepatocellular carcinoma development. PIN1 inhibitor API-1 directly and specifically binds to the Pin1 peptidyl-prolyl isomerase domain and effectively inhibits Pin1 cis-trans isomerizing activity. purity: 98%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.