Learn More
Pigs, Fisher-Free™
SCI_ED
Supplier:  Ranaco Corporation 140.1232.1
 View more versions of this product
Catalog No. S1232S
Add to Cart
Description
Description
Order Info
  • INJECTION KEY
  • Plain: Circulatory system not injected with colors.
  • Single: Arteries injected with red latex.
  • Double: Arteries red, veins blue.
  • Triple: Arteries red, veins blue, and hepatic portal veins yellow.
Specifications
Specifications
Fetal Pig
Double
33.02+ cm
Fisher-Free™
13+ in.
Ea.
Product Suggestions
Product Suggestions