Description
Order Info
- INJECTION KEY
- Plain: Circulatory system not injected with colors.
- Single: Arteries injected with red latex.
- Double: Arteries red, veins blue.
- Triple: Arteries red, veins blue, and hepatic portal veins yellow.
Specifications
Fetal Pig | |
Plain | |
33.02+ cm |
Fisher-Free™ | |
13+ in. | |
5/Cs. |