Learn More
New Pig PIG™ Sediment Drain Filter With Drain Ports
Use this filter bag drain insert to collect sediment, dirt and debris from your catch basin and meet EPA Stormwater Regulations for erosion control and pollution.
Supplier: New Pig FLT749WOPT
This item is not returnable.
View return policy
Description
- Ideal for use in the construction industry and anywhere sediment collection is a concern
- Geotextile constructed of a monofilament polypropylene captures sand, dirt, sediment, trash, leaves, sticks and cigarette butts
- Integrated straps secure the bag in place under the drain grate without the use of rebar and cradle the grate for added support and security during installation and removal
- Straps are high-visibility; easily seen by workers and compliance inspectors
- Can handle large capacity but is manageable during maintenance; no equipment or forklift needed for removal
- Maximum flow rate 3,500 gallons/minute; runoff won't cause flooding when the unit is properly maintained
- Overflow port allows excess water to release into drain during heavy flow or if filter is blocked
Specifications
Polypropylene | |
With Overflow Port | |
1.1 kg |
Drain Cover | |
2.5 lb. |