Learn More
PBL Assay Science VeriKine-HS Pig Interferon Alpha (IFN-Alpha, IFNa) ELISA (High Sensitivity) [1x96 well plate]
Supplier: PBL Assay Science 471001
VeriKine-HS Pig Interferon Alpha ELISA is manufactured by PBL Assay Science (Catalog #47100-1) for research use only (RUO) and packaged as a 1x96-well plate. This assay is serum, plasma, and tissue culture media (TCM) compatible and has an assay range of 2.34-150 pg/mL. This ELISA kit has intra- and inter-assay CVs of less than 10%.