Learn More
New Pig PIG™ Berber Carpet Adhesive-Backed Grippy™ Floor Mat
Keep floors safe and capture dirt and liquid at your door with this carpeted adhesive-backed runner; replace your rental rugs in entrances, walkways and lobbies with this absorbent mat to prevent slips, trips and falls.
Supplier: New Pig GRP913X50BY
This item is not returnable.
View return policy
Description
- Unique PIG Restroom Runner Grippy Mat can be easily rolled out and cut to length for continuous floor coverage with no gaps or overlaps; 25-foot roll is ideal for up to 10 urinals, under rows of sinks or wherever custom sizes are needed
- Permanently infused antimicrobial technology prevents the growth of odor-causing bacteria and mold on the mats to keep restrooms cleaner and fresher smelling
- Proprietary, full-coverage adhesive backing keeps mats in place underfoot with no shifting, curling or flipping up to stop slip and trip hazards; peels up easily
- Mats stick to your floor and won't budge while you clean them; simply mop them with the rest of the floor Durable, Hi-Trac™ Surface stands up to daily cleaning and restroom traffic
- EFM™Absorbent Technology captures urine and water on contact, wicks them away and dries out quickly Impermeable backing layers stop liquid pass-through to prevent stains and grout damage
Specifications
Brown | |
Floor Mat | |
Polyester |