Learn More
Thermo Scientific™ Pierce™ LC-MS/MS System Suitability Standard (7 x 5 mixture)
Pierce LC-MS/MS System Suitability Standard (7 x 5 mixture) contains seven HeavyPeptide AQUA Ultimate peptides provided at five dilutions distinguished by differential isotopic labeling to assess sensitivity and dynamic range of LC-MS/MS systems.
$237.00 - $1132.00
Description
The Pierce LC-MS/MS System Suitability Standard (7 x 5 mixture) contains seven PRTC HeavyPeptide AQUA Ultimate peptides in five isotopologue versions for the generation of seven standard curves to assess sensitivity and dynamic range of LC-MS/MS systems.
The Pierce LC-MS/MS System Suitability Standard (7 x 5 mixture) is:
• High quality—contains five sets of seven HeavyPeptide AQUA Ultimate peptides, each distinguishable through unique isotopologue labeling
• Optimized—enables scheduling of MS acquisition windows for improved quantification and increased multiplexing
• Quantitative—assesses dynamic range of instrument and instrument sensitivity
• Predictive—enables assessment of chromatography and MS instrument performance
Applications:
• Optimization of LC & MS parameters
• Standard for data-dependent acquisition (DDA)
• Standard for targeted MS (e.g., parallel-reaction monitoring (PRM)
• Identification of total peptide elution window
• Assessment dynamic range of nano or capillary flow LC-MS/MS systems
Specifications
LC-MS/MS System Standard | |
Pierce™ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.