Learn More
World Precision Instrument PicoNozzle Kit v1-5430-20
Supplier: World Precision Instrument 543020
The PicoNozzle Kit (version 1) allows micropipettes to be securely mounted in micropositioners for stable axial air delivery. Because air enters the pipette axially, lateral whipping during injection is eliminated.