Learn More
Physitemp PHYSITEMP HEAT LAMP
Supplier: Physitemp HL1
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Heat Lamp, Optional infrared heat lamp is table mounted with a standard U Screw Clamp that expands to fit tables up to 2.25 in. D, An 18 in. adjustable swing arm provides easy adjustment of radiant heat to specimen