Learn More
Detek Inc PHOTOFLO 200 (16OZ)/MIN 5 REQ
Supplier: Detek Inc 1464510
This item has a minimum qty of 6 per supplier requirements.
Photo-Flo 200 Solution; Kodak; 16 oz.; Should be added to the final wash when processing black and white films; Decreases water-surface tension on the emulsion, Minimizes water marks/streaks on film, Promotes faster, more uniform drying